siRNA Details
Gene Symbol: Myt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171615.1 | NP_001165086.1 | 11, 12 | 2037 | ||
NM_001171616.1 | NP_001165087.1 | 10, 11 | 2147 | |||
NM_001171680.1 | NP_001165151.1 | 5, 6 | 2227 | |||
NM_008665.4 | NP_032691.2 | 11, 12 | 2261 | |||
GenBank mRNA | AB082378.1 | - | 11, 12 | 2248 | ||
AB093267.2 | - | 10, 11 | 1854 | |||
AF004294.1 | - | 5, 6 | 2228 | |||
AK133358.1 | - | 5, 6 | 958 | |||
AK135131.1 | - | 3, 4 | 423 | |||
BC063252.1 | - | 11, 12 | 2037 | 6406740 |
