siRNA Details
| Gene Symbol: Ncam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081445.1 | NP_001074914.1 | 7 | 1201 | ||
| NM_001113204.1 | NP_001106675.1 | 7 | 1201 | |||
| NM_010875.3 | NP_035005.2 | 7 | 1201 | |||
| GenBank mRNA | AK051197.1 | - | 7 | 1058 | ||
| AK082621.1 | - | 7 | 1054 | |||
| BC011310.1 | - | 7 | 890 | 3708650 | ||
| BC062942.1 | - | 7 | 1004 | |||
| X15049.1 | - | 7 | 1022 | |||
| Y00051.1 | - | 7 | 1022 | |||

Ncam1
Genomic Map