siRNA Details
| Gene Symbol: Nck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010878.2 | NP_035008.2 | 3, 4 | 1008 | ||
| GenBank mRNA | AF043259.1 | - | 3, 4 | 1096 | ||
| AF084183.1 | - | 2, 3 | 931 | |||
| AK027980.1 | - | 1, 2 | 163 | |||
| AK036870.1 | - | 170 | ||||
| AK053155.1 | - | 805 | ||||
| AK054051.1 | - | 2, 3 | 837 | |||
| BC002015.1 | - | 3, 4 | 1008 | 3593441 | ||

Nck1
Genomic Map