siRNA Details
| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB219976.1 | - | 9 | 1298 | ||
| AF000582.1 | - | 11 | 1449 | |||
| AK052117.1 | - | 11 | 1520 | |||
| AK087443.1 | - | 12 | 1528 | |||
| BC053387.1 | - | 11 | 1643 | 30054358 | ||
| U39060.1 | - | 11 | 1501 | |||

Ncoa2
Genomic Map