siRNA Details
| Gene Symbol: Ndrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U52073.1 | - | 5, 6 | 453 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndrg1| Gene Symbol: Ndrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U52073.1 | - | 5, 6 | 453 | ||