siRNA Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010901.2 | NP_035031.2 | 2, 3 | 1528 | ||
| GenBank mRNA | AK039929.1 | - | 2, 3 | 1445 | ||
| AK051602.1 | - | 2, 3 | 1486 | |||
| AK134054.1 | - | 2, 3 | 1447 | |||
| AK147708.1 | - | 2, 3 | 1499 | |||
| AK155087.1 | - | 2, 3 | 1498 | |||
| AK220187.1 | - | 2, 3 | 1300 | |||
| BC120900.1 | - | 2, 3 | 1247 | 40085687 | ||
| BC120901.1 | - | 2, 3 | 1247 | 40085688 | ||
| D85612.1 | - | 2, 3 | 1244 | |||
| EU887626.1 | - | 2, 3 | 1216 | |||
| EU887627.1 | - | 2, 3 | 1216 | |||
| EU887628.1 | - | 2, 3 | 1216 | |||
| EU887629.1 | - | 2, 3 | 1240 | |||
| EU887630.1 | - | 2, 3 | 1240 | |||
| EU887631.1 | - | 2, 3 | 1240 | |||
| U28807.1 | - | 2, 3 | 1207 | |||

Nfatc3
Genomic Map