siRNA Details
| Gene Symbol: Nfkb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177369.1 | NP_001170840.1 | 16 | 1937 | ||
| NM_001177370.1 | NP_001170841.1 | 15 | 1853 | |||
| NM_019408.3 | NP_062281.1 | 15 | 2088 | |||
| GenBank mRNA | AF155372.1 | - | 16 | 2015 | ||
| AF155373.1 | - | 16 | 2052 | |||
| AK134081.1 | - | 16 | 2356 | |||
| AK137680.1 | - | 16 | 1927 | |||
| AK144994.1 | - | 16 | 1930 | |||
| AK145312.1 | - | 15 | 2054 | |||
| AK148160.1 | - | 16 | 1925 | |||
| AK150012.1 | - | 16 | 1916 | |||
| AK153412.1 | - | 11 | 1497 | |||
| AK196178.1 | - | 1 | 28 | |||
| BC027423.1 | - | 15 | 1854 | 3672913 | ||
| DQ020182.1 | - | 15 | 1785 | |||
| DQ020183.1 | - | 14 | 1906 | |||
| DQ020184.1 | - | 13 | 1986 | |||
| DQ020185.1 | - | 15 | 1785 | |||

Nfkb2
Genomic Map