siRNA Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010911.2 | NP_035041.2 | 11 | 1253 | ||
| GenBank mRNA | AJ222660.1 | - | 11 | 1252 | ||
| AK005724.1 | - | 11 | 1243 | |||
| AK075575.1 | - | 11 | 1251 | |||
| AK132838.1 | - | 11 | 1254 | |||
| AK148527.1 | - | 11 | 1254 | |||
| AK159828.1 | - | 11 | 1242 | |||
| AK189862.1 | - | 3 | 241 | |||
| AK191681.1 | - | 3 | 241 | |||
| AK193838.1 | - | 3 | 241 | |||
| AK198789.1 | - | 3 | 241 | |||
| BC010586.1 | - | 11 | 1231 | 3711352 | ||

Nfs1
Genomic Map