siRNA Details
| Gene Symbol: Nkx3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010921.3 | NP_035051.1 | 2 | 413 | ||
| GenBank mRNA | AK137281.1 | - | 2 | 424 | ||
| BC132401.1 | - | 2 | 467 | 40130678 | ||
| BC132427.1 | - | 2 | 467 | 40130704 | ||
| U73460.1 | - | 2 | 400 | |||
| U88542.1 | - | 2 | 413 | |||

Nkx3-1
Genomic Map