siRNA Details
| Gene Symbol: Notch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010928.2 | NP_035058.2 | 34 | 6926 | ||
| GenBank mRNA | BC059256.1 | - | 3 | 1026 | ||
| D32210.1 | - | 34 | 6917 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Notch2| Gene Symbol: Notch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010928.2 | NP_035058.2 | 34 | 6926 | ||
| GenBank mRNA | BC059256.1 | - | 3 | 1026 | ||
| D32210.1 | - | 34 | 6917 | |||