siRNA Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008715.2 | NP_032741.2 | 17, 18 | 3019 | ||
| GenBank mRNA | AK046055.1 | - | 10, 11 | 1604 | ||
| BC059263.1 | - | 17, 18 | 3019 | 6409580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints6| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008715.2 | NP_032741.2 | 17, 18 | 3019 | ||
| GenBank mRNA | AK046055.1 | - | 10, 11 | 1604 | ||
| BC059263.1 | - | 17, 18 | 3019 | 6409580 | ||