siRNA Details
| Gene Symbol: Npas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008719.2 | NP_032745.2 | 16 | 2080 | ||
| GenBank mRNA | BC109166.2 | - | 16 | 2080 | 40048145 | |
| U77969.1 | - | 16 | 2154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npas2| Gene Symbol: Npas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008719.2 | NP_032745.2 | 16 | 2080 | ||
| GenBank mRNA | BC109166.2 | - | 16 | 2080 | 40048145 | |
| U77969.1 | - | 16 | 2154 | |||