siRNA Details
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008719.2 | NP_032745.2 | 2 | 509 | ||
GenBank mRNA | BC109166.2 | - | 2 | 509 | 40048145 | |
U77969.1 | - | 2 | 583 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Npas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008719.2 | NP_032745.2 | 2 | 509 | ||
GenBank mRNA | BC109166.2 | - | 2 | 509 | 40048145 | |
U77969.1 | - | 2 | 583 |