siRNA Details
| Gene Symbol: Slc11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146161.1 | NP_001139633.1 | 12 | 1294 | ||
| NM_008732.2 | NP_032758.2 | 12 | 1294 | |||
| GenBank mRNA | AF029758.1 | - | 12 | 1229 | ||
| AK049856.1 | - | 12 | 1242 | |||
| AK083478.1 | - | 12 | 1240 | |||
| AK148276.1 | - | 12 | 1242 | |||
| AK193963.1 | - | 1 | 13 | |||
| AK205883.1 | - | 1 | 13 | |||
| BC019137.1 | - | 12 | 1268 | 5040976 | ||
| L33415.1 | - | 12 | 1222 | |||

Slc11a2
Genomic Map