siRNA Details
| Gene Symbol: Nrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164226.1 | NP_001157698.1 | 8 | 1182 | ||
| NM_001164227.1 | NP_001157699.1 | 8 | 1182 | |||
| NM_001164228.1 | NP_001157700.1 | 8 | 1179 | |||
| NM_001164229.1 | NP_001157701.1 | 9 | 1256 | |||
| NM_001164230.1 | NP_001157702.1 | 8 | 1179 | |||
| NM_010938.4 | NP_035068.3 | 8 | 1179 | |||
| GenBank mRNA | AF098077.1 | - | 8 | 1114 | ||
| AK014494.1 | - | 8 | 1143 | |||
| AK029034.1 | - | 8 | 1160 | |||
| AK031103.1 | - | 3 | 383 | |||
| AK037697.1 | - | 8 | 1140 | |||
| AK050861.1 | - | 8 | 1096 | |||
| AK082580.1 | - | 8 | 1180 | |||
| AK134483.1 | - | 8 | 1096 | |||
| AK135673.1 | - | 9 | 1259 | |||
| BC005410.1 | - | 7 | 1000 | 3496661 | ||

Nrf1
Genomic Map