siRNA Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008737.2 | NP_032763.2 | 3, 4 | 927 | ||
| GenBank mRNA | AK082992.1 | - | 3, 4 | 980 | ||
| AK135428.1 | - | 3, 4 | 604 | |||
| AK141964.1 | - | 3, 4 | 719 | |||
| AK148372.1 | - | 3, 4 | 903 | |||
| AK159219.1 | - | 3, 4 | 718 | |||
| AK164378.1 | - | 3, 4 | 929 | |||
| BC060129.1 | - | 3, 4 | 707 | 6409596 | ||
| D50086.1 | - | 3, 4 | 787 | |||

Nrp1
Genomic Map