siRNA Details
Gene Symbol: Nrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077403.1 | NP_001070871.1 | 8 | 2072 | ||
NM_001077404.1 | NP_001070872.1 | 8 | 2072 | |||
NM_001077405.1 | NP_001070873.1 | 8 | 2072 | |||
NM_001077406.1 | NP_001070874.1 | 8 | 2072 | |||
NM_001077407.1 | NP_001070875.1 | 8 | 2072 | |||
NM_010939.2 | NP_035069.2 | 8 | 2072 | |||
GenBank mRNA | AF483506.1 | - | 8 | 1184 | ||
AF483507.1 | - | 8 | 1184 | |||
AK149643.1 | - | 8 | 1998 | |||
AK153553.1 | - | 8 | 1461 | |||
BC057028.1 | - | 8 | 1132 | |||
BC098200.1 | - | 8 | 1185 |
