siRNA Details
| Gene Symbol: Nrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077403.1 | NP_001070871.1 | 14 | 3207 | ||
| NM_001077404.1 | NP_001070872.1 | 14 | 3207 | |||
| NM_001077405.1 | NP_001070873.1 | 14 | 3207 | |||
| NM_001077406.1 | NP_001070874.1 | 14 | 3207 | |||
| NM_001077407.1 | NP_001070875.1 | 14 | 3207 | |||
| NM_010939.2 | NP_035069.2 | 14 | 3207 | |||
| GenBank mRNA | AF483506.1 | - | 14 | 2319 | ||
| AF483507.1 | - | 14 | 2319 | |||
| AK141234.1 | - | 4 | 564 | |||
| AK149643.1 | - | 14 | 3134 | |||
| AK153553.1 | - | 14 | 2596 | |||
| AK203742.1 | - | 2 | 149 | |||
| BC029876.1 | - | 6 | 975 | |||
| BC057028.1 | - | 14 | 2267 | |||
| BC098200.1 | - | 14 | 2320 | |||

Nrp2
Genomic Map