siRNA Details
| Gene Symbol: Nrxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020252.3 | NP_064648.3 | 9 | 2627 | ||
| NM_177284.2 | NP_796258.2 | 7 | 2591 | |||
| GenBank mRNA | AB093249.2 | - | 9 | 2627 | ||
| AK133271.1 | - | 4 | 1218 | |||
| AK137407.1 | - | 8 | 2610 | |||
| BC079893.1 | - | 7 | 2522 | |||

Nrxn1
Genomic Map