siRNA Details
| Gene Symbol: Nrxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020252.3 | NP_064648.3 | 18 | 4479 | ||
| NM_177284.2 | NP_796258.2 | 16 | 4443 | |||
| GenBank mRNA | AB093249.2 | - | 18 | 4479 | ||
| AK017578.1 | - | 1 | 44 | |||
| AK137407.1 | - | 17 | 4463 | |||
| BC079893.1 | - | 16 | 4374 | |||

Nrxn1
Genomic Map