siRNA Details
| Gene Symbol: Nrxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020252.3 | NP_064648.3 | 14 | 3776 | ||
| NM_177284.2 | NP_796258.2 | 12 | 3740 | |||
| GenBank mRNA | AB093249.2 | - | 14 | 3776 | ||
| AK133271.1 | - | 9 | 2367 | |||
| AK137407.1 | - | 13 | 3760 | |||
| BC079893.1 | - | 12 | 3671 | |||

Nrxn1
Genomic Map