siRNA Details
| Gene Symbol: Nrxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198587.3 | NP_001185516.2 | 6 | 2284 | ||
| NM_172544.3 | NP_766132.2 | 5 | 1174 | |||
| GenBank mRNA | AK032126.1 | - | 5 | 1166 | ||
| AK078216.1 | - | 7 | 2596 | |||
| AK082055.1 | - | 7 | 2596 | |||
| AK129205.1 | - | 4 | 1047 | |||
| BC060719.1 | - | 7 | 1859 | 6406001 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK016596.1 | - | 303 | Chr. - - [-] on Build | ||

Nrxn3
Genomic Map