siRNA Details
| Gene Symbol: Ntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008744.2 | NP_032770.2 | 4 | 1533 | ||
| GenBank mRNA | AF128865.1 | - | 3 | 1268 | ||
| AI840271.1 | - | 3 | 407 | |||
| BC141294.1 | - | 3 | 1302 | 9055906 | ||
| BE652257.1 | - | 4 | 292 | |||
| U65418.1 | - | 3 | 1249 | |||

Ntn1
Genomic Map