siRNA Details
| Gene Symbol: Ntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008744.2 | NP_032770.2 | 2 | 799 | ||
| GenBank mRNA | AF128865.1 | - | 1 | 534 | ||
| BC141294.1 | - | 1 | 568 | 9055906 | ||
| U65418.1 | - | 1 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ntn1| Gene Symbol: Ntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008744.2 | NP_032770.2 | 2 | 799 | ||
| GenBank mRNA | AF128865.1 | - | 1 | 534 | ||
| BC141294.1 | - | 1 | 568 | 9055906 | ||
| U65418.1 | - | 1 | 515 | |||