siRNA Details
| Gene Symbol: Ntrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008746.5 | NP_032772.3 | 11 | 1393 | ||
| NM_182809.2 | NP_877961.1 | 11 | 1393 | |||
| GenBank mRNA | AF035399.1 | - | 12 | 1488 | ||
| AF035400.1 | - | 11 | 1542 | |||
| AY336094.1 | - | 11 | 1338 | |||
| BC139764.1 | - | 12 | 1405 | 40110345 | ||

Ntrk3
Genomic Map