siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022563.2 | NP_072075.2 | 11 | 1587 | ||
| GenBank mRNA | AK132504.1 | - | 8 | 1086 | ||
| AK205850.1 | - | 1 | 39 | |||
| BC138826.1 | - | 12 | 1833 | 8861848 | ||
| BC138827.1 | - | 12 | 1728 | 8861849 | ||
| G49238.1 | - | 12 | 1894 | |||
| X76505.1 | - | 12 | 1894 | |||

Ddr2
Genomic Map