siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022563.2 | NP_072075.2 | 5 | 663 | ||
| GenBank mRNA | AK132504.1 | - | 2 | 162 | ||
| BC138826.1 | - | 6 | 909 | 8861848 | ||
| BC138827.1 | - | 6 | 804 | 8861849 | ||
| G49238.1 | - | 6 | 970 | |||
| X76505.1 | - | 6 | 970 | |||

Ddr2
Genomic Map