siRNA Details
| Gene Symbol: Dusp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008748.3 | NP_032774.1 | 5 | 1072 | ||
| GenBank mRNA | BC052705.1 | - | 5 | 830 | 5697787 | |
| X95518.1 | - | 4 | 715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp8| Gene Symbol: Dusp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008748.3 | NP_032774.1 | 5 | 1072 | ||
| GenBank mRNA | BC052705.1 | - | 5 | 830 | 5697787 | |
| X95518.1 | - | 4 | 715 | |||