siRNA Details
| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016978.2 | NP_058674.1 | 3 | 542 | ||
| GenBank mRNA | AK128914.1 | - | 3 | 533 | ||
| AK145877.1 | - | 3 | 543 | |||
| AK146413.1 | - | 3 | 487 | |||
| AK168851.1 | - | 3 | 491 | |||
| AK185243.1 | - | 2 | 327 | |||
| AK185430.1 | - | 2 | 326 | |||
| AK187824.1 | - | 1 | 185 | |||
| BC008119.1 | - | 3 | 475 | 3498240 | ||
| X64837.1 | - | 2 | 516 | |||

Oat
Genomic Map