siRNA Details
| Gene Symbol: Oit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010959.2 | NP_035089.1 | 4 | 685 | ||
| GenBank mRNA | AF356506.1 | - | 4 | 632 | ||
| AF517106.1 | - | 3 | 504 | |||
| AF517107.1 | - | 3 | 504 | |||
| AK040570.1 | - | 4 | 686 | |||
| AK050081.1 | - | 4 | 652 | |||
| AY180915.1 | - | 4 | 609 | |||

Oit3
Genomic Map