siRNA Details
| Gene Symbol: Oit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010959.2 | NP_035089.1 | 8, 9 | 1558 | ||
| GenBank mRNA | AF356506.1 | - | 8, 9 | 1505 | ||
| AF517106.1 | - | 7, 8 | 1377 | |||
| AF517107.1 | - | 7, 8 | 1377 | |||
| AK040570.1 | - | 8, 9 | 1559 | |||
| AK050081.1 | - | 8, 9 | 1525 | |||
| AK153440.1 | - | 4, 5 | 800 | |||
| AY180915.1 | - | 8, 9 | 1482 | |||

Oit3
Genomic Map