siRNA Details
| Gene Symbol: Olfr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147027.2 | NP_667238.2 | 1 | 26 | ||
| GenBank mRNA | BC106818.1 | - | 1 | 22 | ||
| BC106819.1 | - | 1 | 26 | 40043407 | ||
| BC106824.1 | - | 1 | 26 | 40043472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr31| Gene Symbol: Olfr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147027.2 | NP_667238.2 | 1 | 26 | ||
| GenBank mRNA | BC106818.1 | - | 1 | 22 | ||
| BC106819.1 | - | 1 | 26 | 40043407 | ||
| BC106824.1 | - | 1 | 26 | 40043472 | ||