siRNA Details
| Gene Symbol: Olfr239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207175.2 | NP_997058.2 | 1 | 745 | ||
| GenBank mRNA | BC127988.1 | - | 1 | 749 | 40135639 | |
| BC134364.1 | - | 1 | 749 | 40135640 | ||
| Gene Symbol: Gm4461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199062.1 | NP_001185991.1 | 1 | 754 | ||
| Gene Symbol: Olfr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010998.2 | NP_035128.2 | 1 | 745 | ||
| GenBank mRNA | AF102532.1 | - | 1 | 556 | ||
More Information
Set Membership: |
|

Olfr239
Genomic Map