siRNA Details
| Gene Symbol: Olfr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010998.2 | NP_035128.2 | 1 | 268 | ||
| GenBank mRNA | AF102532.1 | - | 1 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr55| Gene Symbol: Olfr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010998.2 | NP_035128.2 | 1 | 268 | ||
| GenBank mRNA | AF102532.1 | - | 1 | 79 | ||
Set Membership: |
|