siRNA Details
| Gene Symbol: Orc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025378.2 | NP_001020549.1 | 9 | 822 | ||
| NM_001271526.1 | NP_001258455.1 | 12 | 1347 | |||
| NM_008765.3 | NP_032791.1 | 12 | 1353 | |||
| GenBank mRNA | AB190256.1 | - | 9 | 812 | ||
| AK048408.1 | - | 12 | 1172 | |||
| AK051747.1 | - | 12 | 1182 | |||
| AK133604.1 | - | 12 | 1173 | |||
| AK146281.1 | - | 12 | 1172 | |||
| AK146859.1 | - | 12 | 1167 | |||
| BC015257.1 | - | 12 | 1163 | 4234871 | ||
| U27457.1 | - | 12 | 1172 | |||

Orc2
Genomic Map