siRNA Details
| Gene Symbol: Sqstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290769.1 | NP_001277698.1 | 3 | 473 | ||
| NM_011018.3 | NP_035148.1 | 3 | 473 | |||
| GenBank mRNA | BC006019.1 | - | 3 | 470 | 3487289 | |
| E08146.1 | - | 3 | 431 | |||
| U40930.1 | - | 3 | 443 | |||
| U57413.1 | - | 3 | 435 | |||

Sqstm1
Genomic Map