siRNA Details
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011020.3 | NP_035150.3 | 11 | 1386 | ||
GenBank mRNA | AB001926.1 | - | 11 | 1352 | ||
AK144225.1 | - | 11 | 1423 | |||
AK163459.1 | - | 12 | 1558 | |||
BC012712.1 | - | 11 | 1455 | 4190161 | ||
BC057002.1 | - | 11 | 1386 | 5702602 | ||
BC110662.1 | - | 11 | 1406 | 4511154 | ||
D49482.1 | - | 11 | 1380 | |||
U23921.1 | - | 11 | 1503 |
