siRNA Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016776.2 | NP_058056.2 | 23 | 3152 | ||
| GenBank mRNA | AK083908.1 | - | 23 | 3178 | ||
| AK143469.1 | - | 23 | 3177 | |||
| AK143487.1 | - | 23 | 3186 | |||
| AK149747.1 | - | 18 | 2461 | |||
| AK154016.1 | - | 21 | 3371 | |||
| AK155074.1 | - | 23 | 3163 | |||
| AK166533.1 | - | 12 | 1352 | |||
| BC004078.1 | - | 13 | 1636 | |||
| BC015070.1 | - | 23 | 3153 | |||
| BC048858.1 | - | 23 | 3152 | 5363740 | ||
| BC052889.1 | - | 23 | 3173 | 30057169 | ||
| BC060167.1 | - | 22 | 3248 | |||
| U63648.1 | - | 23 | 3161 | |||

Mybbp1a
Genomic Map