siRNA Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008777.3 | NP_032803.2 | 11 | 1353 | ||
| GenBank mRNA | AK145839.1 | - | 11 | 1218 | ||
| AK149519.1 | - | 11 | 1216 | |||
| BC013458.1 | - | 11 | 1211 | 4223155 | ||
| X51942.1 | - | 11 | 1178 | |||

Pah
Genomic Map