siRNA Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013626.3 | NP_038654.2 | 25, 26 | 3198 | ||
| GenBank mRNA | BC129869.1 | - | 8, 9 | 878 | ||
| U79523.1 | - | 25, 26 | 3086 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pam| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013626.3 | NP_038654.2 | 25, 26 | 3198 | ||
| GenBank mRNA | BC129869.1 | - | 8, 9 | 878 | ||
| U79523.1 | - | 25, 26 | 3086 | |||