siRNA Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013626.3 | NP_038654.2 | 5 | 759 | ||
| GenBank mRNA | AK049423.1 | - | 4 | 420 | ||
| AK051649.1 | - | 5 | 749 | |||
| U79523.1 | - | 5 | 644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pam| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013626.3 | NP_038654.2 | 5 | 759 | ||
| GenBank mRNA | AK049423.1 | - | 4 | 420 | ||
| AK051649.1 | - | 5 | 749 | |||
| U79523.1 | - | 5 | 644 | |||