siRNA Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008779.2 | NP_032805.2 | 6 | 793 | ||
| GenBank mRNA | L01991.1 | - | 902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn3| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008779.2 | NP_032805.2 | 6 | 793 | ||
| GenBank mRNA | L01991.1 | - | 902 | |||