siRNA Details
| Gene Symbol: Pappa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021362.1 | NP_067337.1 | 13 | 3847 | ||
| GenBank mRNA | AF258461.1 | - | 12 | 2708 | ||
| AF260433.1 | - | 1 | 12 | |||
| AF439513.1 | - | 13 | 3242 | |||
| AF439514.1 | - | 14 | 3329 | |||

Pappa
Genomic Map