siRNA Details
Gene Symbol: Pappa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021362.1 | NP_067337.1 | 19 | 4919 | ||
GenBank mRNA | AF258461.1 | - | 18 | 3780 | ||
AF439513.1 | - | 19 | 4314 | |||
AF439514.1 | - | 20 | 4401 | |||
AK075697.1 | - | 7 | 1083 |
