siRNA Details
Gene Symbol: Pax5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008782.2 | NP_032808.1 | 4, 5 | 496 | ||
GenBank mRNA | JX065130.1 | - | 3, 4 | 377 | ||
M97013.1 | - | 4, 5 | 496 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pax5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008782.2 | NP_032808.1 | 4, 5 | 496 | ||
GenBank mRNA | JX065130.1 | - | 3, 4 | 377 | ||
M97013.1 | - | 4, 5 | 496 |