siRNA Details
| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008782.2 | NP_032808.1 | 3, 4 | 443 | ||
| GenBank mRNA | JX065130.1 | - | 2, 3 | 324 | ||
| M97013.1 | - | 3, 4 | 443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax5| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008782.2 | NP_032808.1 | 3, 4 | 443 | ||
| GenBank mRNA | JX065130.1 | - | 2, 3 | 324 | ||
| M97013.1 | - | 3, 4 | 443 | |||