siRNA Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103165.1 | NP_001096635.1 | 2 | 299 | ||
| NM_001103166.1 | NP_001096636.1 | 2 | 299 | |||
| NM_001174073.1 | NP_001167544.1 | 2 | 275 | |||
| NM_011042.2 | NP_035172.2 | 2 | 299 | |||
| GenBank mRNA | AK012297.1 | - | 2 | 147 | ||
| AK161527.1 | - | 2 | 123 | |||
| AK169268.1 | - | 2 | 393 | |||
| BC020478.1 | - | 1 | 44 | |||
| BQ900329.1 | - | 2 | 275 | |||
| EU604548.1 | - | 1 | 54 | |||
| L19661.1 | - | 2 | 186 | |||

Pcbp2
Genomic Map