siRNA Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK148328.1 | - | 6 | 655 | ||
| BC055030.1 | - | 6 | 643 | 5053475 | ||
| L09192.1 | - | 5 | 661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcx| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK148328.1 | - | 6 | 655 | ||
| BC055030.1 | - | 6 | 643 | 5053475 | ||
| L09192.1 | - | 5 | 661 | |||