siRNA Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 10 | 1151 | ||
NM_001286662.1 | NP_001273591.1 | 10 | 1069 | |||
NM_011062.4 | NP_035192.2 | 10 | 1151 | |||
GenBank mRNA | AF079535.1 | - | 10 | 1108 | ||
AF086625.1 | - | 10 | 1117 | |||
AF126294.1 | - | 10 | 1092 | |||
AK147149.1 | - | 10 | 1143 | |||
AK147835.1 | - | 10 | 1126 | |||
AK149162.1 | - | 10 | 1085 | |||
AK149289.1 | - | 10 | 1109 | |||
AK162677.1 | - | 10 | 1111 | |||
AK165564.1 | - | 10 | 1088 | |||
AK188106.1 | - | 2 | 119 | |||
AK205439.1 | - | 1 | 64 | |||
AK219503.1 | - | 2 | 78 | |||
AY062008.1 | - | 10 | 1047 | |||
BC146001.1 | - | 10 | 1088 | 40131304 | ||
BC146003.1 | - | 10 | 1088 | 40131306 |
