siRNA Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080773.2 | NP_001074242.1 | 4 | 545 | ||
| NM_001286662.1 | NP_001273591.1 | 4 | 463 | |||
| NM_011062.4 | NP_035192.2 | 4 | 545 | |||
| GenBank mRNA | AF079535.1 | - | 4 | 502 | ||
| AF086625.1 | - | 4 | 511 | |||
| AF126294.1 | - | 4 | 486 | |||
| AK143283.1 | - | 3 | 771 | |||
| AK147149.1 | - | 4 | 537 | |||
| AK147835.1 | - | 4 | 520 | |||
| AK149162.1 | - | 4 | 478 | |||
| AK149289.1 | - | 4 | 503 | |||
| AK162677.1 | - | 4 | 505 | |||
| AK165564.1 | - | 4 | 482 | |||
| AY062008.1 | - | 4 | 441 | |||
| BC146001.1 | - | 4 | 482 | 40131304 | ||
| BC146003.1 | - | 4 | 482 | 40131306 | ||
| CJ046953.1 | - | 4 | 464 | |||

Pdpk1
Genomic Map